6-Iodo-1,4-benzodioxane 95% remainder mainly 5-isomer
Leverantör: Thermo Scientific
Synonyms: 6-Iodo-2,3-dihydro-1,4-benzodioxine, 1,2-Ethylenedioxy-4-iodobenzene, 6-Iodo-2,3-dihydrobenzo[b][1,4]dioxine
B20127.06 B20127.14
B20127.06EA 1175 SEK
B20127.06 B20127.14
6-Iodo-1,4-benzodioxane 95% remainder mainly 5-isomer
6-Iodo-1,4-benzodioxane
Purity: 95% CAS No.: 57744-67-9 Molecular Formula: C8H7IO2 Formula Weight: 262.05 Boiling Point: 100-102°/0.25mm MDL No.: MFCD00221460
Formel: C₈H₇IO₂
MW: 262,05 g/mol
Kokpunkt: 100…102 °C (0,25 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD00221460
CAS nummer: 57744-67-9
EINECS: 257-817-2