Propyl-3-propoxypyridine-2-carboxylate ≥98.0%

Leverantör: TCI EUROPE

P1196-1G P1196-5G
TCIAP1196-1GEA 1425 SEK
TCIAP1196-1G TCIAP1196-5G
Propyl-3-propoxypyridine-2-carboxylate ≥98.0%
Propyl-3-propoxypyridine-2-carboxylate
Formel: C₁₂H₁₇NO₃
MDL Number: MFCD00191575
CAS nummer: 134319-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e6fe1eb43a46404a7ff2a4241c819dde76ae0dc&vid=2e6fe1eb43a46404a7ff2a4241c819dde76ae0dc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR