1,3-Di-o-tolylguanidine ≥99.0%

Leverantör: TCI EUROPE
Varning

D0953-25G D0953-500G
TCIAD0953-25GEA 215 SEK
TCIAD0953-25G TCIAD0953-500G
1,3-Di-o-tolylguanidine ≥99.0%
1,3-Di-o-tolylguanidine
Formel: C₁₅H₁₇N₃
MW: 239,32 g/mol
MDL Number: MFCD00008513
CAS nummer: 97-39-2
EINECS: 202-577-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bb011b3796d510a3ebd35eb3e1c0ec0b5bcc75f&vid=1bb011b3796d510a3ebd35eb3e1c0ec0b5bcc75f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 174.0 to 178.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR