1,3-Di-o-tolylguanidine ≥99%
Leverantör: Thermo Scientific
Varning
B25127.14 B25127.22 B25127.36
B25127.14EA 600 SEK
B25127.14 B25127.22 B25127.36
1,3-Di-o-tolylguanidine ≥99%
1,3-Di-o-tolylguanidine
Purity: 99% CAS No.: 97-39-2 Molecular Formula: C15H17N3 Formula Weight: 239.32 Melting Point: 176-178° UN No.: 2811 MDL No.: MFCD00008513
Formel: C₁₅H₁₇N₃
MW: 239,32 g/mol
Smältpunkt: 176…178 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008513
CAS nummer: 97-39-2
EINECS: 202-577-6
UN: 2811
ADR: ,