4-Butylbiphenyl ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 4-Butyl-1,1'-Biphenyl

B3888-5G B3888-25G
TCIAB3888-5GEA 215 SEK
TCIAB3888-5G TCIAB3888-25G
4-Butylbiphenyl ≥98.0%
4-Butylbiphenyl
Formel: C₁₆H₁₈
MW: 210,32 g/mol
Kokpunkt: 129°C
Smältpunkt: 16°C
MDL Number: MFCD00100665
CAS nummer: 37909-95-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a18c73142bc79bd96ac8e2bf0353d2d9d8856b77&vid=a18c73142bc79bd96ac8e2bf0353d2d9d8856b77&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR