5-Chlorobenzimidazole ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
C0130-25G
C0130-5G
TCIAC0130-25GEA
5125
SEK
TCIAC0130-25G
TCIAC0130-5G
5-Chlorobenzimidazole ≥98.0% (by GC, titration analysis)
5-Chlorobenzimidazole
Formel:
C₇H₅ClN₂ |
MDL Number:
MFCD17015012 CAS nummer: 4887-82-5 EINECS: 225-503-4 |
|
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 128.0 to 134.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...