3-Propoxypicolinic acid ≥98.0%

Leverantör: TCI
Varning

P1195-100MG P1195-1G
TCIAP1195-100MGEA 790 SEK
TCIAP1195-100MG TCIAP1195-1G
3-Propoxypicolinic acid ≥98.0%
3-Propoxypicolinic acid
Formel: C₉H₁₁NO₃
Smältpunkt: 118°C
MDL Number: MFCD00191574
CAS nummer: 14440-94-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e6243338e8f3f8f83a7d0f74f8a5a0c09074c22&vid=5e6243338e8f3f8f83a7d0f74f8a5a0c09074c22&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR