1-(4-(Trifluoromethyl)phenyl)thiourea ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: N-[4-(Trifluoromethyl)phenyl]thiourea
L12355.03 L12355.06
L12355.03EA 850 SEK
L12355.03 L12355.06
1-(4-(Trifluoromethyl)phenyl)thiourea ≥98%
1-(4-(Trifluoromethyl)phenyl)thiourea
Purity: 98% CAS No.: 1736-72-7 Molecular Formula: C8H7F3N2S Formula Weight: 220.22 Odour: Odourless Melting Point: 142-146° Beilstein Reference: 2695479 UN No.: 2811 MDL No.: MFCD00041189
Formel: C₈H₇F₃N₂S
MW: 220.22 g/mol
Smältpunkt: 142…146 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041189
CAS nummer: 1736-72-7
ADR: 6.1 III