2-Nitroterephthalic acid-4-methyl ester ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-(Methoxycarbonyl)-2-nitrobenzoic acid
H27779.06 H27779.14
H27779.06EA 1900 SEK
H27779.06 H27779.14
2-Nitroterephthalic acid-4-methyl ester ≥97%
2-Nitroterephthalic acid-4-methyl ester
Purity: 97% CAS No.: 55737-66-1 Molecular Formula: C9H7NO6 Formula Weight: 225.17 Melting Point: 132-136° MDL No.: MFCD06203344
Formel: C₉H₇NO₆
MW: 225,17 g/mol
Smältpunkt: 132…136 °C
Storage Temperature: Omgivande
MDL Number: MFCD06203344
CAS nummer: 55737-66-1