tert-Butylacetyl chloride ≥99.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 3,3-Dimethylbutyryl chloride

B1196-25ML B1196-250ML
TCIAB1196-25MLEA 565 SEK
TCIAB1196-25ML TCIAB1196-250ML
tert-Butylacetyl chloride ≥99.0%
tert-Butylacetyl chloride
Formel: C₆H₁₁ClO
MW: 134,61 g/mol
Kokpunkt: 132°C
Flampunkt: 21°C
MDL Number: MFCD00000737
CAS nummer: 7065-46-5
EINECS: 230-356-4
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee4484562913d07b3f462827d3286f6f12e23ca3&vid=ee4484562913d07b3f462827d3286f6f12e23ca3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Red to Green clear liquid
Purity (Argentmetric Titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR