N-Lauroylsarcosine sodium salt hydrate ≥98.0%
Leverantör: TCI EUROPE
Fara
L0019-25G L0019-500G
TCIAL0019-25GEA 335 SEK
TCIAL0019-25G TCIAL0019-500G
N-Lauroylsarcosine sodium salt hydrate ≥98.0%
N-Lauroylsarcosine sodium salt hydrate
Formel: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
Smältpunkt: 46°C
Storage Temperature: Omgivande
MDL Number: MFCD00042728
CAS nummer: 137-16-6
EINECS: 205-281-5
UN: 2811
ADR: 6.1,II
Merck Index: 14,04368
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a2f7f10bb1daa089d95f0cf1135873c20b78a93&vid=c7590e6ce662994ee0cee40099c987308ca0da8c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 7.0 %