L(+)-Isoleucinol ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
L-Isoleucinol, (S)-(+)-Isoleucinol, (2S,3S)-2-Amino-3-methyl-1-pentanol, (2S,3S)-2-Amino-3-methylpentan-1-ol
I0462-1G
I0462-5G
TCIAI0462-1GEA
630
SEK
TCIAI0462-1G
TCIAI0462-5G
L(+)-Isoleucinol ≥97.0% (by GC, titration analysis)
L(+)-Isoleucinol
Formel:
C₆H₁₅NO MW: 117,19 g/mol |
MDL Number:
MFCD00004731 CAS nummer: 24629-25-2 EINECS: 246-371-4 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 40.0 to 44.0 deg-C |
Specific rotation [a]20/D | +1.0 to +4.0 deg (C=1.6, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...