Benzyl phenyl ether ≥97%
Leverantör: Thermo Fisher Scientific
Synonyms: Benzyl Phenyl Ether
B22539.06 B22539.14 B22539.22
B22539.06EA 1175 SEK
B22539.06 B22539.14 B22539.22
Benzyl phenyl ether ≥97%
Benzyl phenyl ether
Purity: 97% CAS No.: 946-80-5 Molecular Formula: C6H5CH2OC6H5 Formula Weight: 184.24 Flash Point: 110°(230°F) Melting Point: 37-40° Boiling Point: 286-287° Beilstein Reference: 2045713 MDL No.: MFCD00020660
Formel: C₁₃H₁₂O
Kokpunkt: 286…287 °C
Smältpunkt: 37…40 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00020660
CAS nummer: 946-80-5