2,2,4-Trimethyl-2-pentanol ≥95.0%

Leverantör: TCI
Varning

T1962-1G T1962-5G
TCIAT1962-1GEA 815 SEK
TCIAT1962-1G TCIAT1962-5G
2,2,4-Trimethyl-2-pentanol ≥95.0%
2,2,4-Trimethyl-2-pentanol
Formel: C₈H₁₈O
MDL Number: MFCD00101611
CAS nummer: 690-37-9
UN: 1987
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6420a30ed943eeb6d180d84f3b8c7964578c8551&vid=6420a30ed943eeb6d180d84f3b8c7964578c8551&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR