1-(4-Chlorophenyl)cyclopropanecarboxylic acid ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid
H50274.03 H50274.06
H50274.03EA 1700 SEK
H50274.03 H50274.06
1-(4-Chlorophenyl)cyclopropanecarboxylic acid ≥99%
1-(4-Chlorophenyl)cyclopropanecarboxylic acid
Purity: 99% CAS No.: 72934-37-3 Molecular Formula: C10H9ClO2 Formula Weight: 196.64 Melting Point: 152-155° MDL No.: MFCD00001289
Formel: C₁₀H₉ClO₂
MW: 196,64 g/mol
Smältpunkt: 152…155 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001289
CAS nummer: 72934-37-3
EINECS: 277-105-5