5-Acetyl-2-bromopyridine ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Acetyl-6-bromopyridine
H33204.03 H33204.06
H33204.03EA 1625 SEK
H33204.03 H33204.06
5-Acetyl-2-bromopyridine ≥97%
5-Acetyl-2-bromopyridine
Purity: 97% CAS No.: 139042-59-4 Molecular Formula: C7H6BrNO Formula Weight: 200.03 Melting Point: 124-128° MDL No.: MFCD04974527
Formel: C₇H₆BrNO
MW: 200,03 g/mol
Smältpunkt: 124…128 °C
Storage Temperature: Omgivande
MDL Number: MFCD04974527
CAS nummer: 139042-59-4