2-Allyloxytetrahydropyran ≥95.0%

Leverantör: TCI
Varning

Synonyms: 2-(Allyloxy)tetrahydro-2H-pyran

A2391-25G A2391-5G
TCIAA2391-25GEA 2850 SEK
TCIAA2391-25G TCIAA2391-5G
2-Allyloxytetrahydropyran ≥95.0%
2-Allyloxytetrahydropyran
Formel: C₈H₁₄O₂
MW: 142.2 g/mol
Flampunkt: 35°C
Storage Temperature: Kylskåp
MDL Number: MFCD00154089
CAS nummer: 4203-49-0
UN: 3271
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27772231d9422e20fa141029072cfc11b71b43e0&vid=27772231d9422e20fa141029072cfc11b71b43e0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR