Cobalt(II) wolframate ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Cobalt(II) tungsten oxide, Cobalt(II) tungstate
40223.09 40223.18 40223.30
40223.09EA 1625 SEK
40223.09 40223.18 40223.30
Cobalt(II) wolframate ≥99%
Cobalt(II) wolframate
Purity: 99% CAS No.: 10101-58-3 Molecular Formula: CoWO4 Formula Weight: 306.78 Physical Form: Powder Density: 8.42 Solubility: Insoluble in water. Soluble in hot concentrated acids MDL No.: MFCD00016034
pigment, drier for enamels, inks, and paints, electronic devices
Formel: CoO₄W
MW: 306.769 g/mol
Densitet: 8.42
Storage Temperature: Omgivande
MDL Number: MFCD00016034
CAS nummer: 10101-58-3
EINECS: 233-254-8