trans-4-(Boc-amino)cyclohexanol ≥98.0% (by GC, total nitrogen)

Leverantör: TCI EUROPE
Varning

Synonyms: tert-Butyl (trans-4-hydroxycyclohexyl)carbamate, trans-4-Aminocyclohexan-1-ol, N-BOC protected

B3564-1G B3564-5G
TCIAB3564-1GEA 365 SEK
TCIAB3564-1G TCIAB3564-5G
trans-4-(Boc-amino)cyclohexanol ≥98.0% (by GC, total nitrogen)
trans-4-(Boc-amino)cyclohexanol
Formel: C₁₁H₂₁NO₃
MW: 215,28 g/mol
MDL Number: MFCD03844613
CAS nummer: 111300-06-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d4c0d27a812f17846d972fa3cda02a3639c1430&vid=5d4c0d27a812f17846d972fa3cda02a3639c1430&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 169.0 to 173.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR