3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 3,5-Bis(trifluoromethyl)-1,2-diaminobenzene
L18180.06 L18180.03
L18180.06EA 7850 SEK
L18180.06 L18180.03
3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%
3,5-Bis(trifluoromethyl)-o-phenylenediamine
Purity: 97% CAS No.: 367-65-7 Molecular Formula: C8H6F6N2 Formula Weight: 244.14 Melting Point: 44-49° Beilstein Reference: 3344332 UN No.: 2811 MDL No.: MFCD01631430
Formel: C₈H₆F₆N₂
MW: 244,14 g/mol
Smältpunkt: 44…49 °C
Storage Temperature: Omgivande
MDL Number: MFCD01631430
CAS nummer: 367-65-7
ADR: 6.1,III