Nordihydroguaiaretic acid ≥97.0%

Leverantör: TCI EUROPE
Varning

D0800-1G D0800-5G
TCIAD0800-1GEA 885 SEK
TCIAD0800-1G TCIAD0800-5G
Nordihydroguaiaretic acid ≥97.0%
Nordihydroguaiaretic acid
Formel: C₁₈H₂₂O₄
MW: 302,37 g/mol
MDL Number: MFCD00002206
CAS nummer: 500-38-9
EINECS: 207-903-0
Merck Index: 14,06693
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f65e61389007f9372d1cbb58066b870d918b7f6&vid=3f65e61389007f9372d1cbb58066b870d918b7f6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 185.0 to 189.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR