Salicylideneaniline ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 2-((Phenylimino)methyl)phenol

S0267-25G S0267-5G
TCIAS0267-25GEA 1025 SEK
TCIAS0267-25G TCIAS0267-5G
Salicylideneaniline ≥98.0% (by GC, titration analysis)
Salicylideneaniline
Formel: C₁₃H₁₁NO
MW: 197.24 g/mol
Kokpunkt: 151°C
MDL Number: MFCD00020066
CAS nummer: 779-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc56904fd1f3886337e889c6891763a5b920e3fb&vid=cc56904fd1f3886337e889c6891763a5b920e3fb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 50.0 to 53.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR