Salicylideneaniline ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-((Phenylimino)methyl)phenol
B25332.09 B25332.18
B25332.09EA 1550 SEK
B25332.09 B25332.18
Salicylideneaniline ≥98%
Salicylideneaniline
Purity: 98% CAS No.: 779-84-0 Molecular Formula: C13H11NO Formula Weight: 197.24 Melting Point: 48-52° Boiling Point: 151°/2mm MDL No.: MFCD00020066
Formel: C₁₃H₁₁NO
MW: 197.24 g/mol
Kokpunkt: 151 °C (2 mmHg)
Smältpunkt: 48…52 °C
Storage Temperature: Omgivande
MDL Number: MFCD00020066
CAS nummer: 779-84-0