2-(Phenylsulphonyl)aniline ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
A1857-25G
A1857-500G
TCIAA1857-25GEA
1200
SEK
TCIAA1857-25G
TCIAA1857-500G
2-(Phenylsulphonyl)aniline ≥98.0% (by HPLC, total nitrogen)
2-(Phenylsulphonyl)aniline
Formel:
C₁₂H₁₁NO₂S MW: 233,2862 g/mol |
MDL Number:
MFCD00007706 CAS nummer: 4273-98-7 EINECS: 224-271-1 |
Specifikation testresultat
Appearance | White to Light red to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 118.0 to 122.0 deg-C |
Solubility in Methanol | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...