4,4'-Diaminoazobenzene 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4,4'-Azodianiline
401580050 401580250
ACRO401580050EA 5100 SEK
ACRO401580050 ACRO401580250
4,4'-Diaminoazobenzene 95%
4,4'-Diaminoazobenzene
Formel: C₁₂H₁₂N₄
MW: 212.26 g/mol
Smältpunkt: 245 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041892
CAS nummer: 538-41-0
EINECS: 208-690-7
Order Now
Specifikation testresultat
Appearance Brown Powder
Infrared spectrum Conforms
Titration with HClO4 ≥94 %
GC ≥98.5 %