4-Amino-3-(trifluoromethoxy)benzonitrile

Leverantör: Thermo Scientific

TL00680DA TL00680EA
MAYBTL00680DAEA 0 SEK
MAYBTL00680DA MAYBTL00680EA
4-Amino-3-(trifluoromethoxy)benzonitrile
4-Amino-3-(trifluoromethoxy)benzonitrile
Formel: C₈H₅F₃N₂O
MW: 202.1300 g/mol
MDL Number: MFCD00204177
CAS nummer: 175278-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR