4-(Chlorosulphonyl)benzoic acid 96%
Leverantör: Thermo Scientific
Fara
405060250 405060050
ACRO405060250EA 4675 SEK
ACRO405060250 ACRO405060050
4-(Chlorosulphonyl)benzoic acid 96%
4-(Chlorosulphonyl)benzoic acid
Formel: C₇H₅ClO₄S
MW: 220,63 g/mol
Smältpunkt: 230 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007448
CAS nummer: 10130-89-9
EINECS: 233-367-2
UN: 3261
ADR: 8,III
Order Now
Specifikation testresultat
Appearance White to beige Powder or crystals
Infrared spectrum Conforms
Titration excess NaOH ≥95 %
Color scale ≤10 APHA
Free amine ≤0.01 %