1-Phenyl-1-(trimethylsilyloxy)ethylene ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: Trimethyl[(1-phenylvinyl)oxy]silane, 1-Phenyl-1-trimethylsiloxyethylene
L05308.03 L05308.06
L05308.03EA 585 SEK
L05308.03 L05308.06
1-Phenyl-1-(trimethylsilyloxy)ethylene ≥97%
1-Phenyl-1-(trimethylsilyloxy)ethylene
Purity: 97% CAS No.: 13735-81-4 Molecular Formula: C11H16OSi Formula Weight: 192.34 Flash Point: 78°(172°F) Refractive Index: 1.5020 Boiling Point: 53-54°/1mm Beilstein Reference: 1306914 MDL No.: MFCD00008582
Formel: C₁₁H₁₆OSi
MW: 192,34 g/mol
Kokpunkt: 53…54 °C (1 mmHg)
Densitet: 0,938
Flampunkt: 78 °C (172 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00008582
CAS nummer: 13735-81-4
EINECS: 237-308-1