1-Tritylimidazole-4-carbaldehyde ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Trityl-1H-imidazole-4-carboxaldehyde, 1-Trityl-1H-imidazole-4-carbaldehyde, 1-Tritylimidazole-4-carboxaldehyde
B25272.06 B25272.14
B25272.06EA 3275 SEK
B25272.06 B25272.14
1-Tritylimidazole-4-carbaldehyde ≥98%
1-Tritylimidazole-4-carbaldehyde
Purity: 98% CAS No.: 33016-47-6 Molecular Formula: C23H18N2O Formula Weight: 338.41 Melting Point: ca 192° dec. MDL No.: MFCD02179554
Formel: C₂₃H₁₈N₂O
MW: 338,41 g/mol
Smältpunkt: ∼ 192 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD02179554
CAS nummer: 33016-47-6