7-(Trifluoromethyl)isatin ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

T2457-1G T2457-5G
TCIAT2457-1GEA 230 SEK
TCIAT2457-1G TCIAT2457-5G
7-(Trifluoromethyl)isatin ≥98.0% (by GC, titration analysis)
7-(Trifluoromethyl)isatin
Formel: C₉H₄F₃NO₂
MDL Number: MFCD01248408
CAS nummer: 391-12-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cc7277ce4aedc119c3b7a4377e9eeec4485f0d5&vid=5cc7277ce4aedc119c3b7a4377e9eeec4485f0d5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 190.0 to 196.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR