5-Amino-2-mercaptobenzimidazole ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-Amino-1H-benzoimidazole-2-thiol
A1482-25G
A1482-5G
TCIAA1482-25GEA
2225
SEK
TCIAA1482-25G
TCIAA1482-5G
5-Amino-2-mercaptobenzimidazole ≥97.0%
5-Amino-2-mercaptobenzimidazole
Formel:
C₇H₇N₃S MW: 165,22 g/mol |
MDL Number:
MFCD00022671 CAS nummer: 2818-66-8 EINECS: 220-574-8 |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...