4-(4-Fluorobenzoyl)piperidine hydrochloride ≥98%
Leverantör: Thermo Fisher Scientific
Synonyms: (4-Fluorophenyl)(piperidin-4-yl)methanone hydrochloride
B23142.06 B23142.03
B23142.06EA 4025 SEK
B23142.06 B23142.03
4-(4-Fluorobenzoyl)piperidine hydrochloride ≥98%
4-(4-Fluorobenzoyl)piperidine hydrochloride
Purity: 98% CAS No.: 25519-78-2 Molecular Formula: C12H14FNO.HCl Formula Weight: 243.71 MDL No.: MFCD00044912
Formel: C₁₂H₁₅ClFNO
MW: 243.71 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00044912
CAS nummer: 25519-78-2
EINECS: 247-070-0