3-(2-Aminoethylamino)propylmethyldimethoxysilane ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 3-(2-Aminoethylamino)propyldimethoxymethylsilane

A0876-25ML A0876-500ML
TCIAA0876-25MLEA 230 SEK
TCIAA0876-25ML TCIAA0876-500ML
3-(2-Aminoethylamino)propylmethyldimethoxysilane ≥97.0% (by GC, titration analysis)
3-(2-Aminoethylamino)propylmethyldimethoxysilane
Formel: C₈H₂₂N₂O₂Si
MW: 206,36 g/mol
Kokpunkt: 139°C
Flampunkt: 126°C
MDL Number: MFCD00054776
CAS nummer: 3069-29-2
EINECS: 221-336-6
UN: 3267
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c2f525239931652c873b4b0b0c2d28f09d34da7&vid=2c2f525239931652c873b4b0b0c2d28f09d34da7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR