4-(Morpholinosulphonyl)aniline

Leverantör: Thermo Scientific

TL00826DA TL00826EA
MAYBTL00826DAEA 0 SEK
MAYBTL00826DA MAYBTL00826EA
4-(Morpholinosulphonyl)aniline
4-(Morpholinosulphonyl)aniline
Formel: C₁₀H₁₄N₂O₃S
MW: 242,300 g/mol
MDL Number: MFCD00277213
CAS nummer: 21626-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR