1-Boc-4-iodopiperidine 97%
Leverantör: Thermo Fisher Scientific
Varning
437270050 437270010
ACRO437270050EA 4175 SEK
ACRO437270050 ACRO437270010
1-Boc-4-iodopiperidine 97%
1-Boc-4-iodopiperidine
Formel: C₁₀H₁₈INO₂
Smältpunkt: 44…49 °C
Storage Temperature: Kylskåp
MDL Number: MFCD04115041
CAS nummer: 301673-14-3
Order Now
Specifikation testresultat
Appearance White to light brown Crystalline powder or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %