1-Boc-piperidine-3-carboxylic acid ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
N-Boc-DL-nipecotic acid, 1-[(tert-butoxy)carbonyl]piperidine-3-carboxylic acid, 1-Boc-Nipecotic acid, Boc-Nip-OH, 1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid, N-BOC-3-Piperidinecarboxylic acid
B3376-25G
B3376-5G
TCIAB3376-25GEA
1600
SEK
TCIAB3376-25G
TCIAB3376-5G
1-Boc-piperidine-3-carboxylic acid ≥97.0% (by GC, titration analysis)
1-Boc-piperidine-3-carboxylic acid
Formel:
C₁₁H₁₉NO₄ MW: 229,3 g/mol Smältpunkt: 160°C |
MDL Number:
MFCD00673775 CAS nummer: 84358-12-3 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...