Tetrazolium violet, Sigma-Aldrich®
Leverantör: SIGMA ALDRICH MICROSCOPY
M-Clarity™ quality level = MQ200
T0138-10G T0138-1G T0138-5G
T0138-10GEA 5210 SEK
T0138-10G T0138-1G T0138-5G
Tetrazolium violet, Sigma-Aldrich®
Tetrazolium violet
Tetrazolium Violet is a monotetrazolium salt which forms water-insoluble formazans. These salts are mainly used as indicators of extracellular redox activity and cell redox potential. Tetrazolium violet is positively charged and cell-permeable.
Tetrazolium Violet has been used to stain agar layers in bacteriophage plaque formation assays.
Formel: C₂₃H₁₇ClN₄
MW: 384.87 g/mol
Smältpunkt: 247...249 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00011875
CAS nummer: 1719-71-7
EINECS: 217-008-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e16a6b1347ffde93ec758cd40475c39afc584e9&vid=3a968c0fc5f775bda6ad95de98fc1b6d251c5efe&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Carbon 69.5 - 73.2 %
Nitrogen 14.1 - 14.9 %
Purity (TLC) ≥98 %
Solubility (Colour) Light Yellow to Green-Yellow
Solubility (Turbidity)
1 mg/mL, MeOH
Clear