2-(1-Naphthylmethyl)-2-imidazoline hydrochloride 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Naphazoline Hydrochloride, Albalon
179660250 179661000
ACRO179660250EA 925 SEK
ACRO179660250 ACRO179661000
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride 99%
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride
Formel: C₁₄H₁₅ClN₂
MW: 246.738 g/mol
Smältpunkt: 254…260 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012554
CAS nummer: 550-99-2
EINECS: 208-989-2
UN: 2811
ADR: 6.1 II
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
GC ≥97.5 %