Victoria Blue B

Leverantör: CHROMA

Synonyms: Basic blue 26, N-(4-((4-(Dimethylamino)phenyl)(4-(phenylamino)naphthalen-1-yl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride

1B-391.100 1B-391.25
CHRO1B-391.100EA 496.4 SEK
CHRO1B-391.100 CHRO1B-391.25
Victoria Blue B
Victoria blue B
Formel: C₃₃H₃₂ClN₃
MW: 506,803 g/mol
MDL Number: MFCD00011878
CAS nummer: 2580-56-5
EINECS: 219-943-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR