1,4'-Bipiperidine ≥98.0%
Leverantör: TCI
Varning
Synonyms: 4-Piperidinopiperidine
P1638-1G P1638-25G P1638-5G
TCIAP1638-1GEA 485 SEK
TCIAP1638-1G TCIAP1638-25G TCIAP1638-5G
1,4'-Bipiperidine ≥98.0%
1,4'-Bipiperidine
Formel: C₁₀H₂₀N₂
MW: 168.28 g/mol
MDL Number: MFCD00006475
CAS nummer: 4897-50-1
EINECS: 225-522-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caa29e4983da7a528455c45b12a32f0592d3c619&vid=caa29e4983da7a528455c45b12a32f0592d3c619&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 63.0 to 67.0 deg-C