1,4'-Bipiperidine ≥99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-Piperidinopiperidine
B22028.03 B22028.06 B22028.14
B22028.03EA 955 SEK
B22028.03 B22028.06 B22028.14
1,4'-Bipiperidine ≥99%
1,4'-Bipiperidine
Purity: 99% CAS No.: 4897-50-1 Molecular Formula: C10H20N2 Formula Weight: 168.28 Melting Point: 66-70° UN No.: 3259 MDL No.: MFCD00006475
Formel: C₁₀H₂₀N₂
MW: 168.28 g/mol
Smältpunkt: 66…70 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006475
CAS nummer: 4897-50-1
EINECS: 225-522-8
ADR: 8 III