N(α)-Fmoc-L-tryptophan
Leverantör: NOVABIOCHEM
Synonyms: Fmoc-Trp-OH
8.52207.0100 8.52207.0025 8.52207.0005
04-12-1035-100EA 3100 SEK
04-12-1035-100 04-12-1035-25 04-12-1035-5
N(α)-Fmoc-L-tryptophan
N(α)-Fmoc-L-tryptophan
Formel: C₂₆H₂₂N₂O₄
MW: 426.5 g/mol
MDL Number: MFCD00037126
CAS nummer: 35737-15-6
EINECS: 252-706-5
Order Now
SPECIFIKATIONER
Environmentally Preferable