2-Thioxanthine ≥80.0%

Leverantör: TCI

T0225-1G T0225-5G
TCIAT0225-1GEA 815 SEK
TCIAT0225-1G TCIAT0225-5G
2-Thioxanthine ≥80.0%
2-Thioxanthine
Formel: C₅H₄N₄OS
MDL Number: MFCD00031505
CAS nummer: 2487-40-3
EINECS: 219-636-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3572b321ec7b8e7ab4cfa6db28b42fbb1e197c6e&vid=3572b321ec7b8e7ab4cfa6db28b42fbb1e197c6e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (HPLC) min. 80.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR