S-Furfuryl thioacetate ≥97.0%
Leverantör: TCI
|
Varning
|
T1283-10G
T1283-25G
TCIAT1283-10GEA
405
SEK
TCIAT1283-10G
TCIAT1283-25G
S-Furfuryl thioacetate ≥97.0%
S-Furfuryl thioacetate
Formel:
C₇H₈O₂S Kokpunkt: 91°C Flampunkt: 92°C |
MDL Number:
MFCD00010083 CAS nummer: 13678-68-7 EINECS: 237-173-9 |
|
Specifikation testresultat
Appearance | Colorless to Brown clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...