4-Hydroxytetrahydropyran ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Tetrahydro-2H-pyran-4-ol, Tetrahydro-4H-pyran-4-ol

T2825-1G T2825-5G
TCIAT2825-1GEA 550 SEK
TCIAT2825-1G TCIAT2825-5G
4-Hydroxytetrahydropyran ≥97.0%
4-Hydroxytetrahydropyran
Formel: C₅H₁₀O₂
MW: 102,1317 g/mol
Kokpunkt: 190°C
Flampunkt: 88°C
MDL Number: MFCD00006633
CAS nummer: 2081-44-9
EINECS: 218-210-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d27d06aec82f8d2cdcbfba3dd29d920392f29b2&vid=8d27d06aec82f8d2cdcbfba3dd29d920392f29b2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR