4-(Methylthio)benzyl alcohol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 4-(Methylthio)benzyl Alcohol

M2314-25G M2314-5G
TCIAM2314-25GEA 6000 SEK
TCIAM2314-25G TCIAM2314-5G
4-(Methylthio)benzyl alcohol ≥98.0%
4-(Methylthio)benzyl alcohol
Formel: C₈H₁₀OS
MW: 154,23 g/mol
Kokpunkt: 109°C
MDL Number: MFCD00009706
CAS nummer: 3446-90-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b58fb769585375b34fa905a246c305437aa166bc&vid=b58fb769585375b34fa905a246c305437aa166bc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR