2-Cyano-4-fluorophenylboronic acid pinacol ester 96%

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: 2-Cyano-4-fluorophenylboronic Acid Pinacol Ester, 2-Cyano-4-fluorobenzeneboronic acid pinacol ester

H62729.MD
H62729.MDEA 1550 SEK
H62729.MD
2-Cyano-4-fluorophenylboronic acid pinacol ester 96%
2-Cyano-4-fluorophenylboronic acid pinacol ester

Purity: 96% CAS No.: 461451-63-8 Molecular Formula: C13H15BFNO2 Formula Weight: 247.07 Physical Form: Solid

Formel: C₁₃H₁₅BFNO₂
MW: 247.07 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09260444
CAS nummer: 461451-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR