1H-Benzimidazole-5-carboxylic acid 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1H-Benzo[d]imidazole-5-carboxylic acid, Benzimidazole-5-carboxylic acid, 5-Benzimidazolecarboxylic acid
290870250 290870050
ACRO290870250EA 2250 SEK
ACRO290870250 ACRO290870050
1H-Benzimidazole-5-carboxylic acid 98%
1H-Benzimidazole-5-carboxylic acid
Formel: C₈H₆N₂O₂
MW: 162,15 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011555
CAS nummer: 15788-16-6
Order Now
Specifikation testresultat
Appearance Grey to brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %