2,2,2-Trifluoroethyl trifluoromethanesulphonate 95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2,2,2-Trifluoroethyltrifluoromethanesulphonate
H61603.06 H61603.14
H61603.06EA 2600 SEK
H61603.06 H61603.14
2,2,2-Trifluoroethyl trifluoromethanesulphonate 95%
2,2,2-Trifluoroethyl trifluoromethanesulphonate
Formel: C₃H₂F₆O₃S
MW: 232.10 g/mol
Densitet: 1.611
Flampunkt: >110 °C (230 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00671579
CAS nummer: 6226-25-1
UN: 3265
ADR: 8 II