Litiumaluminiumhydrid 4.0 M i Dietyleter, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Aluminium lithium hydride, Lithium tetrahydridoaluminate
199511000 199518000
199511000.EA 1325 SEK
199511000. 199518000.
Litiumaluminiumhydrid 4.0 M i Dietyleter, AcroSeal®
Litiumaluminiumhydrid
Formel: H₄AlLi
MW: 37.95 g/mol
Smältpunkt: 125
Flampunkt: –017 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011075
CAS nummer: 16853-85-3
EINECS: 240-877-9
UN: 1411
ADR: 4.3 I
Merck Index: 13,00348
Order Now
Specifikation testresultat
Appearance Colorless Solution (May develop some turbidity or precipitate)
Titration Iodometric 18.0 to 22.0 % (LiAlH4)